Learn More
OHAUS™ Cable Kit for Dot Matrix Impact Printer
Catalog No. 01919312 Shop All OHAUS Products
Change view
Click to view available options
Printer Cable STP103
Catalog No.
01-919-312 Printer Cable STP103
1 options
Catalog No. 01-919-312 Supplier OHAUS™ Supplier No. 80252573
For use with Trooper™ Industrial Bench Scales.
  • Includes RS232 cable and adapter
  • To connect Dot Matrix Impact Printer to Trooper series scales
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) MB Series, Discovery, Ranger, Defender DxxP
For Use With (Application) Connects Dot Matrix Impact Printer to Trooper seri
Includes RS232 cable and adapter
Type Cable Kit