Learn More
Research Products International Corp OHAUS Endeavor 5000 Light Duty
SDP
Supplier:  Research Products International Corp OH30770911
Encompass
Endeavor(TM) Light Duty Shakers are available in 3 models: 3 mm orbit, 20 mm orbit, or dedicated microplate model. All models can be used in incubators and cold rooms. Stable, accurate, long-term shaking is ensured with the triple eccentric drive and maintenance-free brushless DC motor. Integrated overload and unbalanced detection to protect samples. Budget-Friendly: No need to buy a separate tray. Ready to attach a wide variety of accessories to customize for low to mid volume applications. Programmable: Save time and guarantee reproducible results. Simple on-board menu to set, store, and recall multi-step programs. Data logging and control: Remote function permits PC control and data transfer.
Catalog No. 50-297-9370