Learn More
Research Products International Corp OHAUS Pioneer Precision Balance, Model PX163
SDP
Catalog No. 502114128
Encompass
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. The PX is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point.
Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
PX features a second line display for additional information/guidance, & USB/RS232 connectivity for easy communication.
Unit is not returnable without original packaging box and material. There are no exceptions.
Catalog No. 50-211-4128 Supplier Research Products International Corp Supplier No. OH30429839
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. The PX is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
The PX offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point.
Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX is durably constructed for versatile, long-term use.
PX features a second line display for additional information/guidance, & USB/RS232 connectivity for easy communication.
Unit is not returnable without original packaging box and material. There are no exceptions.