Learn More
Research Products International Corp OHAUS Pioneer Semi-Micro, PX225D
SDP
Supplier:  Research Products International Corp OH30475734
Encompass
The Pioneer PX5 combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. The PX5 is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
The PX5 offers high accuracy and repeatability for essential weighing applications in laboratory, industrial and education settings at an economical price point.
Featuring a cast metal lower housing, sub-pan and stainless steel weighing pan, the PX5 is durably constructed for versatile, long-term use.
The PX5 features a second line display for additional information or guidance, a static removal bar for convenient grounding, and USB and RS232 connectivity.
Unit is not returnable without original packaging box and material. There are no exceptions.
Catalog No. 50-211-4205
Add to Cart
Request a Quote