Learn More
Research Products International Corp OHAUS PR Series Analytical, PR64/E
SDP
Supplier:  Research Products International Corp OH30430060
Encompass
Offering accuracy and repeatability in essential weighing applications in laboratory, industrial and education settings, PR Analytical and Precision Balances deliver competitive performance at an economical price. Featuring RS232 connectivity for easy communication, and a backlit display and a simple interface for uncomplicated operation, the PR is perfectly designed for your workplace.
The PR is equipped with 3 essential weighing modes, RS-232 connectivity for data transfer and storage, and internal calibration (in select models), making it ideal for routine weighing applications.
Equipped with an easy-to-read, bright backlit display and a simple user interface, the PR is incredibly easy to operate, with almost no training required.
The PR's small footprint saves desktop space while providing a large weighing surface.
Unit is not returnable without original packaging box and material. There are no exceptions.
Catalog No. 50-211-4206