Learn More
OHAUS™ RS232 Cable and Adapter for 8025042 Printers
Catalog No. 02211381 Shop All OHAUS Products
Change view
Click to view available options
Cable and Adapter for 8025042 Impact Printer
Catalog No.
02-211-381 Cable and Adapter for 8025042 Impact Printer
1 options
Catalog No. 02-211-381 Supplier OHAUS™ Supplier No. 80500570
Connects the AP, Voyager, Voyager Pro, Explorer, Explorer Pro, and Adventurer balances to Compact Dot Matrix Impact Printers (8025042).
For use with AP, Voyager, Voyager Pro, Explorer, Explorer Pro, and Adventurer balances.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) AP, Voyager, Voyager Pro, Explorer, Explorer
For Use With (Application) Connects the AP, Voyager, Voyager Pro, Explorer, E
Languages Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX)
Product Line Printer Cables
Type RS232 Cable and Adapter