Learn More
Research Products International Corp OHAUS Scout SPX, SPX8200
SDP
Catalog No. 502114179
Encompass
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Unit is not returnable without original packaging box and material. There are no exceptions.
Catalog No. 50-211-4179 Supplier Research Products International Corp Supplier No. OH30253028
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Unit is not returnable without original packaging box and material. There are no exceptions.