Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Starter Water Analysis Portable pH Meters
SCI_ED

Catalog No. S05079 Shop All OHAUS Products
Change view
Click to view available options
Model:
ST300
ST300-B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
S05079 ST300-B
S05080 ST300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S05079 Supplier OHAUS™ Supplier No. 83033962/EMD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LCD meters combine portability with precise pH measurement.

  • Auto buffer recognition stores calibration data to avoid errors
  • Automatic/manual temperature compensation for accurate readings
  • ORP or temperature electrode connectivity for additional functionality
  • IP54 protection shields water/dust damage
  • Size: 90 x 150 x 35mm
  • Weight: 0.18kg
TRUSTED_SUSTAINABILITY

Specifications

Includes 4 AAA Batteries
Accuracy (Temperature) ±0.5°C
Accuracy (mV) ±1
Accuracy (pH) ±0.01
Data Management Details 30 Sets, most recent calibration data
Measurement Modes pH with mV and Temperature
Calibration Automatic/Manual
Range (pH) 0 to 14
Resolution (pH) 0.01
Range (mV) -1999 to 1999
Resolution (mV) 1mV
Calibration Points 1 to 3
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Backlight Option None
Data Points 30
Log Function Type 1000 Measurement, 10 Calibration
Inputs BNC, NTC
IP Rating IP54
Weight (Metric) 0.18 kg
Material ABS
Model ST300-B
Resolution 0.01pH; 1mV; 0.1°C
Battery Type AAA
Type Portable pH Meters
Battery Life >500 hr.
Width (English) 3.5 in.
Width (Metric) 9 cm
Height (English) 1.3 in.
Height (Metric) 3.5 cm
Depth (Metric) 17 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.