Learn More
Cole-Parmer Portable Balance 2200g x 0.01g with Touchscreen
Supplier:  Cole-Parmer 1101517
Encompass
Ohaus STX2202 ScoutPortable Balance 2200 g x 0.01 g with Touchscreen, Superior performance and easy operationRemovable high strength stainless steel weighing platformImpact-resistant pan supportIntegrated transportation/storage lockSealed front panel and molded spill ring help keep the Scout safe from spillsDesigned to be stackable when utilizing the stacking and storage cover (optional on the SPX)
Catalog No. NC1606138