Learn More
Research Products International Corp OHAUS Temperature Probe 20 cm PTFE
SDP
Catalog No. 502230204
Encompass
PTFE coated temperature probe for digital hotplates and hotplate stirrers, probe length 20 cm, temperature range of 40 - 200C
Catalog No. 50-223-0204 Supplier Research Products International Corp Supplier No. OH30500591
PTFE coated temperature probe for digital hotplates and hotplate stirrers, probe length 20 cm, temperature range of 40 - 200C