Learn More
Research Products International Corp OHAUS Temperature Probe 20 cm S/S
SDP
Catalog No. 502230205
Encompass
Stainless steel temperature probe for digital hotplates and hotplate stirrers. Probe length 20 cm, temperature range of 40 - 400C
Catalog No. 50-223-0205 Supplier Research Products International Corp Supplier No. OH30500590
Stainless steel temperature probe for digital hotplates and hotplate stirrers. Probe length 20 cm, temperature range of 40 - 400C