Learn More
Sigma Aldrich Fine Chemicals Biosciences OhausR ExplorerR precision1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7605281EA
Encompass
Explorer delivers accurate results within seconds improving operator efficiency productivity and throughput with a stabilization time up to 50% faster. The frameless flip-top design provides unobstructed access to the weighing chamber.The expansive side entry 160 mm x 240 mm allows you to freely place and remove large weigh boats or other large vessels in the weighing chamber. Antistatic coated glass helps dissipate static charges in the weighing chamber that could adversely affect the weighing results..nbspIntuitive Software Easy-to-use graphical software featuring 14 applications virtual QWERTY and numeric keypads and below-minimum weight indication.nbspDraftshield Explorers draftshield provides ample access and visibility to the weighing chamber and features antistatic coated glass.nbspModular Design Explorers modular design features a color touch display that can be separated from the weighing base.nbspUSB and RS232 and optional third port of either RS232 or Ethernet.nbspStai
Catalog No. 50-290-8908