Learn More
Sigma Aldrich Fine Chemicals Biosciences OhausR ExplorerR precision1EA
Catalog No. 502908889
Encompass
Display5.7 (145 mm) Full-color VGA graphic touchscreen display with user-controlled brightnessOperationAC adapter (included)CommunicationRS232 and USB (included)ConstructionModular design metal base ABS top housing stainless steel pan glass draftshield with top mounted side doors and flip/sliding top door (1mg models only) illuminated up-front level indicator integral weigh below hook security bracket calibration lock four touchless sensors full housing in-use coverDesign FeaturesSelectable environmental filters auto tare user selectable span calibration points software lockout and reset menu user selectable communication settings and data print options user definable project and user IDs auto standby up to 14 operating languages
Catalog No. 50-290-8889 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7439221EA
Display5.7 (145 mm) Full-color VGA graphic touchscreen display with user-controlled brightnessOperationAC adapter (included)CommunicationRS232 and USB (included)ConstructionModular design metal base ABS top housing stainless steel pan glass draftshield with top mounted side doors and flip/sliding top door (1mg models only) illuminated up-front level indicator integral weigh below hook security bracket calibration lock four touchless sensors full housing in-use coverDesign FeaturesSelectable environmental filters auto tare user selectable span calibration points software lockout and reset menu user selectable communication settings and data print options user definable project and user IDs auto standby up to 14 operating languages