Learn More
Sigma Aldrich Fine Chemicals Biosciences OhausR ScoutR SPX Balance1EA
Catalog No. 502908566
Encompass
Ideal for laboratory and industrial applications the OHAUS Scout comes in a slim stackable design with large backlit LCD. Features include superior overload protection multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high reSOLUTIONon weighing results these portable balances set a new standard in laboratory and industrial weighing.
Catalog No. 50-290-8566 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7422241EA
Ideal for laboratory and industrial applications the OHAUS Scout comes in a slim stackable design with large backlit LCD. Features include superior overload protection multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high reSOLUTIONon weighing results these portable balances set a new standard in laboratory and industrial weighing.