Learn More
Bellco Glass, Inc. Over Head Drive Drive Vessel,36L Six Vertical Sidearms
SDP
Catalog No. 501944793
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. Compatible with bellco over head drive and magnetic stirrers. MADE IN THE USA. Specifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 0Sidearms: 6OD x OAH: 410 x 535mm
Catalog No. 50-194-4793 Supplier Bellco Glass, Inc. Supplier No. 196436000
Manufactured from type 1 class a Borosilicate glass. Includes 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. Compatible with bellco over head drive and magnetic stirrers. MADE IN THE USA. Specifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 0Sidearms: 6OD x OAH: 410 x 535mm