Learn More
Bellco Glass, Inc. Over Head Drive PTFE Impeller Assembly ,15L
SDP
Catalog No. 501944785
Encompass_Preferred
Ideal use for over head drive vessels. 15L OHD Impeller Assembly Consists of Paddle, Plug, Paddle Holder, andDrive Shaft. MADE IN THE USAPaddle [A523-214]: PTFE material, 171.5mm wide x 76.2mm tall x 1.6mm thickPlug [A542-49]: PTFE, 9.5mm OD x 19.1mm long (approx.)Paddle Holder [A542-50]: White PTFE, 279.4mm long x 19.1mm ODDrive Shaft [A542-47]: 316L SST, 215.9mm long x 6.4mm OD (approx.)
Catalog No. 50-194-4785 Supplier Bellco Glass, Inc. Supplier No. 196430015
Ideal use for over head drive vessels. 15L OHD Impeller Assembly Consists of Paddle, Plug, Paddle Holder, andDrive Shaft. MADE IN THE USAPaddle [A523-214]: PTFE material, 171.5mm wide x 76.2mm tall x 1.6mm thickPlug [A542-49]: PTFE, 9.5mm OD x 19.1mm long (approx.)Paddle Holder [A542-50]: White PTFE, 279.4mm long x 19.1mm ODDrive Shaft [A542-47]: 316L SST, 215.9mm long x 6.4mm OD (approx.)