Learn More
Bellco Glass, Inc. Over Head Drive PTFE Impeller Assembly , 8L
SDP
Catalog No. 501944784
Encompass_Preferred
Ideal use for over head drive vessels. Description: 8L OHD Impeller Assembly Consists of Paddle, Plug, Paddle Holder, andDrive Shaft. MADE IN THE USAPaddle [A523-214]: PTFE material, 171.5mm wide x 76.2mm tall x 1.6mm thickPlug [A542-49]: PTFE, 9.5mm OD x 19.1mm long (approx.)Paddle Holder [A542-50]: White PTFE, 279.4mm long x 19.1mm ODDrive Shaft [A542-46]: 316L SST, 190.5mm long x 6.4mm OD (approx.)
Catalog No. 50-194-4784 Supplier Bellco Glass, Inc. Supplier No. 196430008
Ideal use for over head drive vessels. Description: 8L OHD Impeller Assembly Consists of Paddle, Plug, Paddle Holder, andDrive Shaft. MADE IN THE USAPaddle [A523-214]: PTFE material, 171.5mm wide x 76.2mm tall x 1.6mm thickPlug [A542-49]: PTFE, 9.5mm OD x 19.1mm long (approx.)Paddle Holder [A542-50]: White PTFE, 279.4mm long x 19.1mm ODDrive Shaft [A542-46]: 316L SST, 190.5mm long x 6.4mm OD (approx.)