Learn More
Bellco Glass, Inc. Over Head Drive Vessel w/6 Side Arms,15L No Baffles
SDP
Catalog No. 501944773
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Includes 4 or 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. MADE IN THE USA. Specifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 0Sidearms: 6OD x OAH: 295 x 460mm
Catalog No. 50-194-4773 Supplier Bellco Glass, Inc. Supplier No. 196415060
Manufactured from type 1 class a Borosilicate glass. Includes 4 or 6 45mm polypropylene blue screw caps and pouring rings, also 1 100mm polypropylene blue cap for the center. Suitable for all cell culture fermentation. MADE IN THE USA. Specifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 0Sidearms: 6OD x OAH: 295 x 460mm