Learn More
Bellco Glass, Inc. Over Head Drive Vessel w/6 Side Arms,8L
SDP
Catalog No. 501944765
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate glass. Includes 4 or 6 polypropylene blue screw caps 45mm and pouring rings, also 1 polypropylene blue cap 100mm for the center. MADE IN THE USASpecifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 0Sidearms: 6OD x OAH: 240 x 430mm
Catalog No. 50-194-4765 Supplier Bellco Glass, Inc. Supplier No. 196408060
Manufactured from Type 1 Class A Borosilicate glass. Includes 4 or 6 polypropylene blue screw caps 45mm and pouring rings, also 1 polypropylene blue cap 100mm for the center. MADE IN THE USASpecifications:Material: Type 1 Class A Borosilicate Glass100mm Center NeckBaffles: 0Sidearms: 6OD x OAH: 240 x 430mm