Learn More
AdipoGen Ohira-Bestmann Reagent
SDP
Supplier:  AdipoGen CDXD0501G001
Encompass_Preferred
Chemical. CAS 90965-06-3. Formula C5H9N2O4P. MW 192.11. Synthetic. Used in the Seyferth-Gilbert homologation which is a chemical reaction of an aryl ketone or aldehyde with dimethyl- or diazomethyl-phosphonate and potassium tert-butoxide to give substituted alkynes.
Catalog No. 50-256-1068