Learn More
MOLECULAR DEPOT LLC OHp-g-cyclodextrin - 25 mL
Catalog No. 5000013452
Encompass_Preferred
10perc 2-OHp-g-cyclodextrin is an aqueous solution of the cyclic hydroxylated dextrin of type Gamma This product has been used as versatile tools in molecular assemblies enhancement of solubility and organic synthesis It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only
Catalog No. 50-000-13452 Supplier MOLECULAR DEPOT LLC Supplier No. K1010005E
10perc 2-OHp-g-cyclodextrin is an aqueous solution of the cyclic hydroxylated dextrin of type Gamma This product has been used as versatile tools in molecular assemblies enhancement of solubility and organic synthesis It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only