Learn More
Chemglass Life Sciences OHS 20 Digital Overhead Stirrer, 115V/60Hz
SDP
Catalog No. 501656622
Encompass_Preferred
Laboratory overhead stirrer with Digital control. Stirrer features a bright WHITE LED display for accurate speed settings. Stirrer features torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Suitable for light mixing task with a maximum viscosity of 10,000 mPa/centipoise.
Catalog No. 50-165-6622 Supplier Chemglass Life Sciences Supplier No. CG2051020
Laboratory overhead stirrer with Digital control. Stirrer features a bright WHITE LED display for accurate speed settings. Stirrer features torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Suitable for light mixing task with a maximum viscosity of 10,000 mPa/centipoise.