Learn More
Chemglass Life Sciences OHS 200 Advance Overhead Stirrer, 115V/60Hz
SDP
Catalog No. 501656629
Encompass_Preferred
Wi-Fi Enabled Laboratory overhead stirrer with a TFT 3.5¿ Display that shows all setting information at a glance. Wi-Fi Connection to Velp Ermes Platform allows the end user monitor status and data logging on the cloud. Stirrer features torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Suitable for mixing task with a maximum viscosity of 100,000 mPa/centipoise.
Catalog No. 50-165-6629 Supplier Chemglass Life Sciences Supplier No. CG2054200
Add to Cart
Add to Cart
Wi-Fi Enabled Laboratory overhead stirrer with a TFT 3.5¿ Display that shows all setting information at a glance. Wi-Fi Connection to Velp Ermes Platform allows the end user monitor status and data logging on the cloud. Stirrer features torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Suitable for mixing task with a maximum viscosity of 100,000 mPa/centipoise.