Learn More
Chemglass Life Sciences OHS 40 Digital Overhead Stirrer, 115V/60Hz
SDP
Catalog No. 501656623
Encompass_Preferred
Laboratory overhead stirrer with Digital control. Stirrer features a bright WHITE LED display for accurate speed settings. Stirrer features torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Suitable for mixing task with a maximum viscosity of 25,000 mPa/centipoise.
Catalog No. 50-165-6623 Supplier Chemglass Life Sciences Supplier No. CG2051040
Laboratory overhead stirrer with Digital control. Stirrer features a bright WHITE LED display for accurate speed settings. Stirrer features torque compensation so the stirrer maintains a constant speed under varying viscosities. Display shows set and actual speed, torque trend and timer. Suitable for mixing task with a maximum viscosity of 25,000 mPa/centipoise.