Learn More
Bel-Art™ Oil-Based Tech Pen
Catalog No. 13245184 Shop All SP Bel-Art & SP Wilmad Products
Change view
Click to view available options
Tip Style:
Soft Tip
Catalog No. Tip Style
13-245-184 Soft Tip
1 options
Catalog No. 13-245-184 Supplier Bel-Art™ Supplier No. F133841000
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Heavy-duty soft tip disposable pens come in a multipack of 7 colors and writes on glass, metal, ceramics, plastics, paper, etc.
  • Oil-based marking paint resists washing, weathering, mild acids and alkalis
  • Operates within the temperature range of -46°C to +65°C (-50°F to +150°F)
  • Withstands autoclaving at 121°C (250°F)
  • Mark is approximately 1 mm (0.040 in.) wide
Specifications
Product Type Permanent Marking Pen
Tip Style Soft Tip
Color Multi
Sterility Non-sterile
Disposable Limited-use
Latex Free Yes
No. per Pack 7
Width (Metric) Line 1 mm