Learn More
Chemglass Life Sciences Oil Bath Combination Package #2
SDP
Catalog No. CG1100200
Encompass_Preferred
Oil Bath Combination Package #2. PLEASE NOTE: HOT PLATE MUST BE PURCHASED SEPARATELY. Package #2 / CG-1100-200, Consists of One Each:CG-1100-02; 150 x 75mm Oil BathCG-15001-30; Digital Temp Controller with ThermocoupleCG-1100-12; Thermocouple ClipCG-1100-31; 1/2 Gallon Bath Oil
Catalog No. CG1100200 Supplier Chemglass Life Sciences Supplier No. CG1100200
Oil Bath Combination Package #2. PLEASE NOTE: HOT PLATE MUST BE PURCHASED SEPARATELY. Package #2 / CG-1100-200, Consists of One Each:CG-1100-02; 150 x 75mm Oil BathCG-15001-30; Digital Temp Controller with ThermocoupleCG-1100-12; Thermocouple ClipCG-1100-31; 1/2 Gallon Bath Oil