Learn More
Chemglass Life Sciences Oil Bath Fluid, Quart
SDP
Catalog No. 50871932
Encompass_Preferred
Oil Bath Fluid, Quart. High temperature silicone bath fluid for use with CG-1100 series oils bath dishes.Viscosity (@25 Deg C) 100cst Operating Temperatures, Deg C -40 to 315 Flashpoint Deg C (Open Cup) >288° Pour Point (Deg C) -65 Coefficient of Expansion 0.000095 cc/cc/DegC Specific Heat, Cal/gm/Deg C 34 Thermal Conductivity, gm- 0.00026 cal/s-cm2/Deg C
Catalog No. 50-871-932 Supplier Chemglass Life Sciences Supplier No. CG110030
Oil Bath Fluid, Quart. High temperature silicone bath fluid for use with CG-1100 series oils bath dishes.Viscosity (@25 Deg C) 100cst Operating Temperatures, Deg C -40 to 315 Flashpoint Deg C (Open Cup) >288° Pour Point (Deg C) -65 Coefficient of Expansion 0.000095 cc/cc/DegC Specific Heat, Cal/gm/Deg C 34 Thermal Conductivity, gm- 0.00026 cal/s-cm2/Deg C