Learn More
Chemglass Life Sciences Oil Bath, 950mL, 150 x 75mm, 120V, 800W
SDP
Catalog No. NC1420925
Encompass_Preferred
Our new low form oil bath is constructed completely of stainless steel and provides for rapid heating of flasks, beakers, etc. Heating rates run between 10-15 per minute with excellent thermal response. Since the bath is constructed entirely of non-magnetic stainless steel, the contents of the vessel along with the oil itself are able to be magnetically stirred. Each bath is supplied complete with a stainless steel lid to keep the oil clean during storage. Although all sizes of the baths are fitted with a 120V heater, a temperature controller such as the CG-15001 is required to maintain the desired heating temperature. Unit is equipped with a permanently attached 13 inch, 3-wire grounded cord having stainless steel overbraid and a female Twist-Lock connection. Each bath comes complete with a 6 foot grounded extension cord with Twist-Lock connection.
Catalog No. NC1420925 Supplier Chemglass Life Sciences Supplier No. CG110002
Our new low form oil bath is constructed completely of stainless steel and provides for rapid heating of flasks, beakers, etc. Heating rates run between 10-15 per minute with excellent thermal response. Since the bath is constructed entirely of non-magnetic stainless steel, the contents of the vessel along with the oil itself are able to be magnetically stirred. Each bath is supplied complete with a stainless steel lid to keep the oil clean during storage. Although all sizes of the baths are fitted with a 120V heater, a temperature controller such as the CG-15001 is required to maintain the desired heating temperature. Unit is equipped with a permanently attached 13 inch, 3-wire grounded cord having stainless steel overbraid and a female Twist-Lock connection. Each bath comes complete with a 6 foot grounded extension cord with Twist-Lock connection.