Learn More
Thermo Scientific OIL/CORROSION PROTECTION
Supplier:  Thermo Scientific 70009824
Encompass_Preferred
anti-corrisive oil to be used with Thermo Scientific Sorvall centrifuges and Fiberlite rotors.
Catalog No. NC9182614