Learn More
Chemglass Life Sciences Chemglass Airfree Oil Diffusion Pump With Bypass, Left Handed (AF-0460-01) - for AF-0450 manifold and 100 mL heating mantle
SDP
Catalog No. 502733605
Encompass_Preferred
The Chemglass Airfree oil diffusion pump with bypass (AF-0460-01) is a single-stage Schlenk-style diffusion pump designed for laboratory vacuum systems. It features a round-bottom boiler intended for use with a 100 mL heating mantle and includes a bypass to protect the pump oil during roughing and start-up.
  • Single-stage diffusion pump design for high vacuum applications.
  • Round-bottom boiler compatible with a 100 mL heating mantle.
  • Bypass isolates the diffusion pump from the mechanical roughing pump during start-up.
  • Achieves vacuum on the order of 10^-5 torr.
  • Pumping rate approximately 8 L/sec.
  • Tubular connections: 1 in OD for heavy-wall vacuum tubing or compatible connectors.
  • Available in left-handed and right-handed orientations.
  • Valves sized for 0-12 mm tubing.
Catalog No. 50-273-3605 Supplier Chemglass Life Sciences Supplier No. AF046001
Add to Cart
Add to Cart
The Chemglass Airfree oil diffusion pump with bypass (AF-0460-01) is a single-stage Schlenk-style diffusion pump designed for laboratory vacuum systems. It features a round-bottom boiler intended for use with a 100 mL heating mantle and includes a bypass to protect the pump oil during roughing and start-up.
  • Single-stage diffusion pump design for high vacuum applications.
  • Round-bottom boiler compatible with a 100 mL heating mantle.
  • Bypass isolates the diffusion pump from the mechanical roughing pump during start-up.
  • Achieves vacuum on the order of 10^-5 torr.
  • Pumping rate approximately 8 L/sec.
  • Tubular connections: 1 in OD for heavy-wall vacuum tubing or compatible connectors.
  • Available in left-handed and right-handed orientations.
  • Valves sized for 0-12 mm tubing.