Learn More
MSC Oil-Dri Absorbent, Granular, General Absorbent, Fullers Earth, 35 lb
Catalog No. 5000794177
Encompass
Oil-Dri Absorbent is a 35 lb bag of granular, general absorbent designed to absorb a wide range of fluids. It is effective for automotive fluids, coolants, fuel, grease, ink, lubricants, oil, oil-based liquids, paint, and water.
  • More absorbent pound for pound
  • Lighter weight clay, easier to handle
  • Reduced dust in each bag
  • More bags per pallet equals lower shipping costs per bag
Catalog No. 50-007-94177 Supplier MSC Supplier No. 43439355
Oil-Dri Absorbent is a 35 lb bag of granular, general absorbent designed to absorb a wide range of fluids. It is effective for automotive fluids, coolants, fuel, grease, ink, lubricants, oil, oil-based liquids, paint, and water.
  • More absorbent pound for pound
  • Lighter weight clay, easier to handle
  • Reduced dust in each bag
  • More bags per pallet equals lower shipping costs per bag