Learn More
MSC Simpson Pump Oil, 15W40, Non-foaming, Pressure Washer
Catalog No. 502838998
Encompass
This premium pump crankcase oil is a proprietary 15W40 blend, engineered to maximize pump performance and protect critical components. Its non-foaming formula ensures compatibility with most pressure washer pumps, preventing wear and corrosion.
  • Proprietary 15W40 blend.
  • Non-foaming formula.
  • Maximizes pump performance.
  • Protects critical components by preventing wear and corrosion.
  • Compatible with most pressure washer pumps.
  • Recommended oil change after initial 50 hours, then every 100 hours or every 3 months.
  • Supplied in a 1 U.S. Pint (16 ounces) container.
Catalog No. 50-283-8998 Supplier MSC Supplier No. 81435992
Add to Cart
Add to Cart
Request a Quote
This premium pump crankcase oil is a proprietary 15W40 blend, engineered to maximize pump performance and protect critical components. Its non-foaming formula ensures compatibility with most pressure washer pumps, preventing wear and corrosion.
  • Proprietary 15W40 blend.
  • Non-foaming formula.
  • Maximizes pump performance.
  • Protects critical components by preventing wear and corrosion.
  • Compatible with most pressure washer pumps.
  • Recommended oil change after initial 50 hours, then every 100 hours or every 3 months.
  • Supplied in a 1 U.S. Pint (16 ounces) container.