Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum Dry Scroll Pump, HiScroll 12 C-Version, Corrosive Gas, DN 25 ISO-KF
Catalog No. NC3978970
Encompass_Preferred
The Pfeiffer HiScroll C series is a dry scroll pump for light corrosive gases. It offers safe, oil-free operation with simplified installation due to its non-return valve and hermetically sealed pumping system.
  • Corrosive gas ready with FFKM check valves, DLC-coated components, and stainless steel flanges.
  • High quality, steady pumping, and short cycle times.
  • Low noise and vibration levels.
  • Over 50% lower energy consumption.
  • Dry, oil-free operation for clean vacuum.
  • Safe operation via non-return valve and hermetically sealed pumping system.
  • Short maintenance times and long intervals.
  • Compact, hermetically sealed design with best-in-class pump-down time.
  • Optimized temperature regulation.
  • Integrates with control systems via RS-485, ProfiNet or EtherCAT interfaces.
Catalog No. NC3978970 Supplier PFEIFFER VACUUM INC Supplier No. 2000223066
The Pfeiffer HiScroll C series is a dry scroll pump for light corrosive gases. It offers safe, oil-free operation with simplified installation due to its non-return valve and hermetically sealed pumping system.
  • Corrosive gas ready with FFKM check valves, DLC-coated components, and stainless steel flanges.
  • High quality, steady pumping, and short cycle times.
  • Low noise and vibration levels.
  • Over 50% lower energy consumption.
  • Dry, oil-free operation for clean vacuum.
  • Safe operation via non-return valve and hermetically sealed pumping system.
  • Short maintenance times and long intervals.
  • Compact, hermetically sealed design with best-in-class pump-down time.
  • Optimized temperature regulation.
  • Integrates with control systems via RS-485, ProfiNet or EtherCAT interfaces.