Learn More
PFEIFFER VACUUM INC Pfeiffer Vacuum HiScroll Dry Scroll Pump, C-Version, Corrosive, 18.1 m³/h, Manual Gas Ballast
Catalog No. NC3978972
Encompass_Preferred
The HiScroll 18 C-Version Corrosive Dry Scroll Pump is an oil-free, high-performance pump designed for demanding applications involving corrosive gases. It offers steady pumping speed, low noise levels, and reduced energy consumption.
  • Corrosive gas ready with FFKM check valves, DLC-coated components, and stainless steel flanges.
  • High quality performance with steady pumping speed and short cycle times.
  • Low noise levels and lower energy consumption.
  • Dry and oil-free pumping principle.
  • Safe operation via standard non-return valve and hermetically sealed pumping system.
  • Simple installation, setup, and short maintenance intervals for low operating costs.
  • Integrates with external systems via RS-485 or ProfiNet/EtherCAT interfaces.
  • Suitable for chemical processing, mass spectrometry, and general laboratory applications.
Catalog No. NC3978972 Supplier PFEIFFER VACUUM INC Supplier No. 2000223067
The HiScroll 18 C-Version Corrosive Dry Scroll Pump is an oil-free, high-performance pump designed for demanding applications involving corrosive gases. It offers steady pumping speed, low noise levels, and reduced energy consumption.
  • Corrosive gas ready with FFKM check valves, DLC-coated components, and stainless steel flanges.
  • High quality performance with steady pumping speed and short cycle times.
  • Low noise levels and lower energy consumption.
  • Dry and oil-free pumping principle.
  • Safe operation via standard non-return valve and hermetically sealed pumping system.
  • Simple installation, setup, and short maintenance intervals for low operating costs.
  • Integrates with external systems via RS-485 or ProfiNet/EtherCAT interfaces.
  • Suitable for chemical processing, mass spectrometry, and general laboratory applications.