Learn More
PFEIFFER VACUUM INC Oil-Free Scroll Vacuum Pump, HiScroll, Pfeiffer Vacuum Inc., Hiscroll 6, Ultimate Vacuum: 1.5E-2 mbar, Pumping Speed: 3.6 CFM, Controller: Omni Control, Internal Gauge: RPT (Pirani)
Catalog No. NC3978956
Encompass_Preferred
Oil-Free Scroll Vacuum Pump, HiScroll, Pfeiffer Vacuum Inc., Hiscroll 6, Ultimate Vacuum: 1.5E-2 mbar, Pumping Speed: 3.6 CFM, Controller: Omni Control, Internal Gauge: RPT (Pirani), Gauge Interface Cable: Included, Connections: Clamping Ring & O-Ring Included, Electrical: Mains Cable Included, Dust Filter: Included, Noise Level: 48 dB(A), Weight: 19kg, Dimensions (L x W x H): 380 x 220 x 257 mm, DN 25 ISO-KF/DN 25 ISO-KF
Catalog No. NC3978956 Supplier PFEIFFER VACUUM INC Supplier No. 2000026531
Oil-Free Scroll Vacuum Pump, HiScroll, Pfeiffer Vacuum Inc., Hiscroll 6, Ultimate Vacuum: 1.5E-2 mbar, Pumping Speed: 3.6 CFM, Controller: Omni Control, Internal Gauge: RPT (Pirani), Gauge Interface Cable: Included, Connections: Clamping Ring & O-Ring Included, Electrical: Mains Cable Included, Dust Filter: Included, Noise Level: 48 dB(A), Weight: 19kg, Dimensions (L x W x H): 380 x 220 x 257 mm, DN 25 ISO-KF/DN 25 ISO-KF