Learn More
Biotage Oil and Grease Standards 20/PK
SDP
Catalog No. NC9948039
Encompass
Oil and Grease Standards are convenient one time use directly from the teflon-PFA tube. Snip open the tube, pour all of the solution into the extraction vessel, and proceed with analysis. Each snip contains exactly 10ml of 20mg n-Hexadecane and 20mg Stearic Acid. Dissolved in 99.5%acetone. Lot certified. Certificate of Analysis
Catalog No. NC9948039 Supplier Biotage Supplier No. 50021HT
Oil and Grease Standards are convenient one time use directly from the teflon-PFA tube. Snip open the tube, pour all of the solution into the extraction vessel, and proceed with analysis. Each snip contains exactly 10ml of 20mg n-Hexadecane and 20mg Stearic Acid. Dissolved in 99.5%acetone. Lot certified. Certificate of Analysis