Learn More
Biotage OIL GREASE STD
SDP
Catalog No. NC9864827
Encompass
Oil & Grease Standards in multi use glass vials. Simply pipete 5 ml of solution into the extraction vessel. Each vial contains 26 ml of 40 mg Stearic Acid and n-Hexadecane solution, for a total of 5 uses per vial.
Catalog No. NC9864827 Supplier Biotage Supplier No. 50003HT
Oil & Grease Standards in multi use glass vials. Simply pipete 5 ml of solution into the extraction vessel. Each vial contains 26 ml of 40 mg Stearic Acid and n-Hexadecane solution, for a total of 5 uses per vial.