Learn More
Oil and Grease Wastewater Standard (ULTRAcheck™ Blind QC Standard)
Catalog No. USQCI770
Encompass
Change view
Click to view available options
Quantity:
10 mL
Catalog No. Quantity
US-QCI-770 10 mL
1 options
Catalog No. US-QCI-770 Supplier Agilent Technologies Supplier No. QCI770
Total Oil and Grease at 10-100mg/L when diluted to 1 liter
Specifications
Chemical Name or Material Oil and Grease Wastewater Standard
Name Note ULTRAcheck™ Blind QC Standard
Packaging Ampule
Quantity 10 mL