Learn More
Biotage Pacific Premium Oil & Grease Disk, 47 mm
SDP
Catalog No. NC9918377
Encompass
Pacific Premium disks, available in both 47 mm and 90 mm, offer excellent recovery and precision. Pacific Premium Oil and Grease SPE Disks provide fast flow rates with dirty samples while offering high recovery values for the oil and grease fraction of the sample. These disks blend various diameters of high quality borosilicate glass micro-fibers; the upper surface of each disk provides a greater porosity, essential for handling particulate heavy samples, and the bottom layer, comprised of smaller micro-fibers and packing material, retains the oil and grease fraction.
Catalog No. NC9918377 Supplier Biotage Supplier No. 166447PHT
Pacific Premium disks, available in both 47 mm and 90 mm, offer excellent recovery and precision. Pacific Premium Oil and Grease SPE Disks provide fast flow rates with dirty samples while offering high recovery values for the oil and grease fraction of the sample. These disks blend various diameters of high quality borosilicate glass micro-fibers; the upper surface of each disk provides a greater porosity, essential for handling particulate heavy samples, and the bottom layer, comprised of smaller micro-fibers and packing material, retains the oil and grease fraction.