Learn More
Grainger Westward Oil Lube Dispenser, Lever Action, 2.14 oz. Per Stroke, 1 ea.
Catalog No. 502754897
Encompass_Preferred
This lever action oil lube dispenser is designed for efficient fluid transfer, providing an output of 2.14 fl oz per stroke. It is suitable for use with transmission differentials and heavier oils, making it a robust solution for various lubrication tasks.
  • Lever action design for controlled manual pumping
  • Compatible with transmission differentials and heavier oils
  • Output capacity of 2.14 fl oz per stroke
  • Includes a 15 in. drum cover, 30 in. pump, and 4 ft. PVC hose with a steel spout
  • Features a 4 in. spout length and 1/4 in. NPT fluid outlet thread
Catalog No. 50-275-4897 Supplier Grainger Supplier No. 15F213
This lever action oil lube dispenser is designed for efficient fluid transfer, providing an output of 2.14 fl oz per stroke. It is suitable for use with transmission differentials and heavier oils, making it a robust solution for various lubrication tasks.
  • Lever action design for controlled manual pumping
  • Compatible with transmission differentials and heavier oils
  • Output capacity of 2.14 fl oz per stroke
  • Includes a 15 in. drum cover, 30 in. pump, and 4 ft. PVC hose with a steel spout
  • Features a 4 in. spout length and 1/4 in. NPT fluid outlet thread