Learn More
Ethos Biosciences Immersion Oil, Low Viscosity, 2oz, Ethos
Catalog No. 5000574285
Encompass_Preferred
Low Viscosity Immersion Oil is a high-quality optical medium designed for microscopy applications requiring clear, high-resolution imaging. Formulated to match the refractive index of glass, it enhances light transmission and image clarity when used with oil immersion objectives. The low-viscosity formulation allows for easy application, smooth slide movement, and simple cleanup, making it ideal for routine and high-throughput microscopy workflows. Suitable for clinical, research, educational, and industrial laboratories, it provides consistent optical performance and reliable results. Supplied in convenient, chemically compatible containers for precise dispensing and storage.
Catalog No. 50-005-74285 Supplier Ethos Biosciences Supplier No. 551002
Low Viscosity Immersion Oil is a high-quality optical medium designed for microscopy applications requiring clear, high-resolution imaging. Formulated to match the refractive index of glass, it enhances light transmission and image clarity when used with oil immersion objectives. The low-viscosity formulation allows for easy application, smooth slide movement, and simple cleanup, making it ideal for routine and high-throughput microscopy workflows. Suitable for clinical, research, educational, and industrial laboratories, it provides consistent optical performance and reliable results. Supplied in convenient, chemically compatible containers for precise dispensing and storage.