Learn More
Waters Corp Waters Oil Mist Filter, Edwards EMF10, for RV5 Pump
Supplier:  Waters Corp 6060139
Encompass_Preferred
This oil mist filter is designed for use with the Edwards RV5 vacuum pump. It is identified as the Edwards EMF10 Oil Mist Filter and includes an oil return kit. This assembly is fitted to the outlet or exhaust port of the pump, helping to manage oil mist and, for QDa systems, eliminating the need for ballast when the oil return kit is installed.
  • Designed for use with the Edwards RV5 vacuum pump.
  • Functions as an Edwards EMF10 oil mist filter.
  • Includes an oil return kit for efficient operation.
  • Fitted to the pump’s outlet/exhaust port.
  • Eliminates the need for ballast on QDa systems when the oil return kit is installed.
  • Do not use the black NW25 pump adaptor provided in the kit.
Catalog No. 50-792-159