Learn More
Welch™ Oil Mist Separator for CRVpro Direct Drive Rotary Vane Vacuum Pump
Catalog No. 01184159
Change view
Click to view available options
For Use With:
Rotary vane pump
Catalog No. For Use With
01-184-159 Rotary vane pump
1 options
Catalog No. 01-184-159 Supplier Welch™ Supplier No. 320015
Add to Cart
Add to Cart
For use with CRVpro Direct Drive Rotary Vane Vacuum Pump
  • Outlet oil separator removes oil particles from exhaust stream, minimizing oil loss to atmosphere
Specifications
Description Outlet Oil Mist Separator
For Use With (Application) Removes oil particles from exhaust stream
For Use With Rotary vane pump