Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oil of Origanum, TCI America™
SDP

Catalog No. O005125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O005125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O005125G Supplier TCI America Supplier No. O005125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8007-11-2
MDL Number MFCD00131768
Synonym Origanum Oil

Specifications

Quantity 25 g
UN Number 2810
Physical Form Liquid
Chemical Name or Material Oil of Origanum
RTECSNumber RM2660000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.