Learn More
Thermo Scientific Oil Vacuum Pump 3L Each
Supplier:  Thermo Scientific 1094301
Encompass
Thermo Scientific 3L/min rotary vane oil lubricated vacuum pump reaches 10^-3 mbar for vacuum drying, impregnation, embedding, and moderate vacuum processes.
Catalog No. NC0028369