Learn More
Apexbio Technology LLC Oil Red O 1320-06-5 1g
SDP
Catalog No. 5000093985
Encompass_Preferred
Oil Red O (CAS 1320-06-5) is a lipophilic diazo dye that selectively stains neutral lipids and cholesterol esters It is designed to enable visualization and quantification of lipid accumulation in biological specimens Oil Red O exerts its activity by binding to hydrophobic lipid droplets In murine liver biopsy samples Oil Red O staining facilitates detection and quantification of hepatic steatosis Based on these properties Oil Red O holds research potential in fields such as metabolic disorder studies investigations of liver disease progression and analysis of related pathophysiological mechanisms
Catalog No. 50-000-93985 Supplier Apexbio Technology LLC Supplier No. A47151000
Oil Red O (CAS 1320-06-5) is a lipophilic diazo dye that selectively stains neutral lipids and cholesterol esters It is designed to enable visualization and quantification of lipid accumulation in biological specimens Oil Red O exerts its activity by binding to hydrophobic lipid droplets In murine liver biopsy samples Oil Red O staining facilitates detection and quantification of hepatic steatosis Based on these properties Oil Red O holds research potential in fields such as metabolic disorder studies investigations of liver disease progression and analysis of related pathophysiological mechanisms