Learn More
Sigma Aldrich Fine Chemicals Biosciences Oil Red O, 1320-06-5, MFCD00003898, 25 g
Catalog No. NC0961554
Encompass
Empirical Formula (Hill Notation): C26H24N4O, Molecular Weight: 408.49, certified by the Biological Stain Commission, Synonym: 1-([4-(Xylylazo)xylyl]azo)-2-naphthol, 1-[2,5-Dimethyl-4-(2,5-dimethylphenylazo)phenylazo]-2-naphthol, Solvent Red 27. Oil Red O is a lysochrome. It is used to stain and visualise general localization of fats.
Catalog No. NC0961554 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. O062525G
Empirical Formula (Hill Notation): C26H24N4O, Molecular Weight: 408.49, certified by the Biological Stain Commission, Synonym: 1-([4-(Xylylazo)xylyl]azo)-2-naphthol, 1-[2,5-Dimethyl-4-(2,5-dimethylphenylazo)phenylazo]-2-naphthol, Solvent Red 27. Oil Red O is a lysochrome. It is used to stain and visualise general localization of fats.