Learn More
Electron Microscopy Sciences Oil Red O, Certified C.I. DcRo-3 10 GR
SDP
Catalog No. 501901630
Encompass_Preferred
Oil Red O is lysochrome (fat soluble dye) used demonstrate fat or lipids in fresh tissue sections. The polyazo group of dyes include the Oil Red, Sudan Red and the Sudan Blacks - all these dyes are interchangeable. Also known as Solvent Red 27, Sudan Red 5B.
Catalog No. 50-190-1630 Supplier Electron Microscopy Sciences Supplier No. 19056
Oil Red O is lysochrome (fat soluble dye) used demonstrate fat or lipids in fresh tissue sections. The polyazo group of dyes include the Oil Red, Sudan Red and the Sudan Blacks - all these dyes are interchangeable. Also known as Solvent Red 27, Sudan Red 5B.