Learn More
Newcomer Supply Fat, Oil Red O, Propylene Glycol Stain Kit, 250 ml kit
SDP
Catalog No. NC2102270
Encompass
Newcomer Supply Fat, Oil Red O, Propylene Glycol Stain Kit procedure, classified as a physical staining method, is used for identification of fat/lipid in frozen sections.
This item is not returnable. View return policy
Catalog No. NC2102270 Supplier Newcomer Supply Supplier No. 9119A
Newcomer Supply Fat, Oil Red O, Propylene Glycol Stain Kit procedure, classified as a physical staining method, is used for identification of fat/lipid in frozen sections.