Learn More
Electron Microscopy Sciences Oil Red O Solution, 0.5% 500 ML
SDP
Catalog No. 5031862
Encompass_Preferred
Used in Oil Red O Method for Lipofuscin, Pigment stain. See references Luna, L.G., (ED.), manual of Histologic Staining Methods of the AFIP, 3rd Edition. The AFIP, 3rd Edition, McGraw-Hill, NY, p 186, c 1968.
Catalog No. 50-318-62 Supplier Electron Microscopy Sciences Supplier No. 2660901
Used in Oil Red O Method for Lipofuscin, Pigment stain. See references Luna, L.G., (ED.), manual of Histologic Staining Methods of the AFIP, 3rd Edition. The AFIP, 3rd Edition, McGraw-Hill, NY, p 186, c 1968.